Tisk…
Burettes, lateral stopcock, Schellbach
Dodavatel: Technosklo
QUALICOLOR
612-4187EA 1320 CZK
612-4189 612-4188 612-4187 612-4193 612-4194 612-4190
Burettes, lateral stopcock, Schellbach
Byrety
Borosilicate glass, Class AS, QUALICOLOR.
  • Schellbach stripe, ring marks at major graduations
  • Blue graduations
DIIN EN ISO 385
Certifikace: DIIN EN ISO 385