Tisk…
Funnels with short stem
Dodavatel: TECHNOSKLO
632.421001041 632.421001056 632.421001071
221-0255EA 65.6 CZK
221-0255 221-0258 221-0260
Funnels with short stem
Nálevky Nálevky filtrační Nálevky k filtraci
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With smooth internal walls. Angle 60°.