1,2-Dibromo-4,5-difluorobenzene ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
A14804.06
A14804.14
A14804.22
A14804.06EA
1730
CZK
A14804.06
A14804.14
A14804.22
1,2-Dibromo-4,5-difluorobenzene ≥98%
1,2-Dibromo-4,5-difluorobenzene
Purity: 98%
CAS No.: 64695-78-9
Molecular Formula: C6H2Br2F2
Formula Weight: 271.89
Melting Point: 31-35°
Beilstein Reference: 1941132
Vzorec:
C₆H₂Br₂F₂ MW: 271,89 g/mol Bod varu: 208 °C (1013 hPa) Bod tání: 31…35 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009890 CAS číslo: 64695-78-9 EINECS: 265-021-1 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...