2-Bromo-4-chloroaniline ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
L03808.14
L03808.06
L03808.14EA
6630
CZK
L03808.14
L03808.06
2-Bromo-4-chloroaniline ≥98%
2-Bromo-4-chloroaniline
Purity: 98%
CAS No.: 873-38-1
Molecular Formula: C6H5BrClN
Formula Weight: 206.47
Odour: Characteristic
Melting Point: 63-68°
Boiling Point: 127°/20mm
Beilstein Reference: 2802563
UN No.: 2811
MDL No.: MFCD00041313
Vzorec:
C₆H₅BrClN Bod varu: 127 °C (20 mmHg) Bod tání: 63…68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041313 CAS číslo: 873-38-1 EINECS: 212-837-0 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...