Tisk…

1-Bromo-3-chloro-4-fluorobenzene ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 4-Bromo-2-chloro-1-fluorobenzene, 4-Bromo-2-chlorofluorobenzene

A19782.06 A19782.14 A19782.22
A19782.06EA 1410 CZK
A19782.06 A19782.14 A19782.22
1-Bromo-3-chloro-4-fluorobenzene ≥98%
1-Bromo-3-chloro-4-fluorobenzene

Purity: 98+% CAS No.: 60811-21-4 Molecular Formula: C6H3BrClF Formula Weight: 209.45 Flash Point: 88°(190°F) Refractive Index: 1.5540 Boiling Point: 188-190° Beilstein Reference: 2206652 UN No.: 3082 MDL No.: MFCD00051794

Vzorec: C₆H₃BrClF
MW: 209,44 g/mol
Bod varu: 188…190 °C
Hustota: 1,727
Bod vzplanutí: 88 °C (190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051794
CAS číslo: 60811-21-4
EINECS: 405-580-2
UN: 3082
ADR: 9,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR