3-Bromo-5-chloro-2-hydroxybenzaldehyde ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3-Bromo-5-chlorosalicylaldehyde
A13449.06
A13449.14
A13449.06EA
1620
CZK
A13449.06
A13449.14
3-Bromo-5-chloro-2-hydroxybenzaldehyde ≥97%
3-Bromo-5-chloro-2-hydroxybenzaldehyde
Purity: 97%
CAS No.: 19652-32-5
Molecular Formula: C7H4BrClO2
Formula Weight: 235.47
Melting Point: 84-88°
Beilstein Reference: 2362159
Vzorec:
C₇H₄BrClO₂ MW: 235,46 g/mol Bod tání: 84…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051690 CAS číslo: 19652-32-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...