Tisk…

1-Bromo-4-chlorobenzene ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 4-Bromochlorobenzene

A12160.0B A12160.22 A12160.30
A12160.0BEA 3850 CZK
A12160.0B A12160.22 A12160.30
1-Bromo-4-chlorobenzene ≥98%
1-Bromo-4-chlorobenzene

Purity: 98+% CAS No.: 106-39-8 Molecular Formula: C6H4BrCl Formula Weight: 191.46 Flash Point: 100°(212°F) Odour: Aromatic Boiling Point: 196° Beilstein Reference: 1099014 UN No.: 2811 MDL No.: MFCD00000600

Vzorec: C₆H₄BrCl
MW: 191.45
Bod varu: 196 °C
Bod tání: 63…67 °C
Hustota: 1.651
Bod vzplanutí: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000600
CAS číslo: 106-39-8
EINECS: 203-392-3
UN: 2811
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR