1-Bromo-3-chlorobenzene ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3-Bromochlorobenzene
A14026.22
A14026.36
A14026.22EA
1790
CZK
A14026.22
A14026.36
1-Bromo-3-chlorobenzene ≥99%
1-Bromo-3-chlorobenzene
Purity: 99%
CAS No.: 108-37-2
Molecular Formula: C6H4BrCl
Formula Weight: 191.46
Flash Point: 80°(176°F)
Refractive Index: 1.5770
Odour: Aromatic
Melting Point: -22°
Boiling Point: 195-196°
Beilstein Reference: 1446353
MDL No.: MFCD00000568
Vzorec:
C₆H₄BrCl MW: 191,45 g/mol Bod varu: 195…196 °C Bod tání: –22 °C Hustota: 1,630 Bod vzplanutí: 80 °C (176 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000568 CAS číslo: 108-37-2 EINECS: 203-575-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...