Tisk…

2-Bromo-1,4-dichlorobenzene ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 1-Bromo-2,5-dichlorobenzene

A12056.06 A12056.14
A12056.06EA 1100 CZK
A12056.06 A12056.14
2-Bromo-1,4-dichlorobenzene ≥98%
2-Bromo-1,4-dichlorobenzene

Purity: 98% CAS No.: 1435-50-3 Molecular Formula: C6H3BrCl2 Formula Weight: 225.91 Flash Point: 110°(230°F) Melting Point: 32-36° Boiling Point: 118-120°/20mm Beilstein Reference: 1934815 MDL No.: MFCD00018505

Vzorec: C₆H₃BrCl₂
MW: 225,9 g/mol
Bod varu: 118…120 °C (20 mmHg)
Bod tání: 32…36 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00018505
CAS číslo: 1435-50-3
EINECS: 215-859-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR