4-Bromo-3,5-dihydroxybenzoic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A18914.14
A18914.22
A18914.14EA
1830
CZK
A18914.14
A18914.22
4-Bromo-3,5-dihydroxybenzoic acid ≥98%
4-Bromo-3,5-dihydroxybenzoic acid
Purity: 98+%
CAS No.: 16534-12-6
Molecular Formula: C7H5BrO4
Formula Weight: 233.02
Melting Point: 268-272°
Beilstein Reference: 2368265
Vzorec:
C₇H₅BrO₄ MW: 233.02 Bod tání: 268…272 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002513 CAS číslo: 16534-12-6 EINECS: 240-606-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...