Tisk…

3-Bromo-4-methoxybenzoic acid ≥98%

Dodavatel: Thermo Scientific
Varování

A12653.06 A12653.14
A12653.06EA 2560 CZK
A12653.06 A12653.14
3-Bromo-4-methoxybenzoic acid ≥98%
3-Bromo-4-methoxybenzoic acid

Purity: 98+% CAS No.: 99-58-1 Molecular Formula: C8H7BrO3 Formula Weight: 231.05 Odour: Odourless Melting Point: 217-222° Beilstein Reference: 2087585 MDL No.: MFCD00020295

Vzorec: C₈H₇BrO₃
MW: 231,05 g/mol
Bod tání: 217…222 °C
Storage Temperature: Ambient
MDL Number: MFCD00020295
CAS číslo: 99-58-1
EINECS: 202-768-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR