Tisk…
Pouring rings for laboratory bottles
Dodavatel: Lenz Laborglas GmbH & CO.KG
07105600 07105601
LENZ07105600EA 689 CZK
LENZ07105600 LENZ07105601
Pouring rings for laboratory bottles
Uzávěry Uzávěry k láhvím
Pouring rings are made of ETFE.
  • For DIN GL threads
  • Heat resistant up to +180 °C