Tisk…
Pouring rings for laboratory bottles
Dodavatel: Lenz Laborglas GmbH & CO.KG
07105400 07105401
LENZ07105400EA 270 CZK
LENZ07105400 LENZ07105401
Pouring rings for laboratory bottles
Uzávěry Uzávěry k láhvím
Pouring rings, made of polypropylene.
  • For DIN GL threads
  • Heat resistant up to +140 °C
  • Good chemical resistance