4-Bromobenzonitrile ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
p-Bromobenzonitrile
A10914.18
A10914.09
A10914.30
A10914.18EA
5220
CZK
A10914.18
A10914.09
A10914.30
4-Bromobenzonitrile ≥98%
4-Bromobenzonitrile
Purity: 98+%
CAS No.: 623-00-7
Molecular Formula: C7H4BrN
Formula Weight: 182.03
Flash Point: 130°(266°F)
Melting Point: 111-114°
Boiling Point: 235-237°
Beilstein Reference: 2041518
UN No.: 3439
MDL No.: MFCD00001811
Vzorec:
C₇H₄BrN MW: 182,02 g/mol Bod varu: 235…237 °C Bod tání: 111…114 °C Hustota: 1,562 Bod vzplanutí: 130 °C (266 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001811 CAS číslo: 623-00-7 EINECS: 210-764-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...