3-Bromobenzonitrile ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A13317.09
A13317.18
A13317.30
A13317.09EA
1690
CZK
A13317.09
A13317.18
A13317.30
3-Bromobenzonitrile ≥99%
3-Bromobenzonitrile
Purity: 99%
CAS No.: 6952-59-6
Molecular Formula: C7H4BrN
Formula Weight: 182.03
Flash Point: 113°(235°F)
Melting Point: 37-41°
Boiling Point: 224-226°
Beilstein Reference: 1858942
UN No.: 3439
MDL No.: MFCD00001796
Vzorec:
C₇H₄BrN Bod varu: 224…226 °C Bod tání: 37…41 °C Hustota: 1,562 Bod vzplanutí: 113 °C (235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001796 CAS číslo: 6952-59-6 EINECS: 230-127-9 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...