2-Bromobenzonitrile ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
o-Bromobenzonitrile
A10679.06
A10679.14
A10679.22
A10679.06EA
1330
CZK
A10679.06
A10679.14
A10679.22
2-Bromobenzonitrile ≥99%
2-Bromobenzonitrile
Purity: 99%
CAS No.: 2042-37-7
Molecular Formula: C7H4BrN
Formula Weight: 182.03
Flash Point:
Vzorec:
C₇H₄BrN MW: 182,02 g/mol Bod varu: 251…253 °C Bod tání: 53…56 °C Hustota: 1,496 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001772 CAS číslo: 2042-37-7 EINECS: 218-045-1 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...