Tisk…

5,6-Dibromovanillin ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2,3-Dibromo-4-hydroxy-5-methoxybenzaldehyde

B21140.03 B21140.06 B21140.14
B21140.03EA 1130 CZK
B21140.03 B21140.06 B21140.14
5,6-Dibromovanillin ≥98%
5,6-Dibromovanillin

Purity: 98% CAS No.: 2973-75-3 Molecular Formula: C8H6Br2O3 Formula Weight: 309.94 Melting Point: 213-217° Beilstein Reference: 1968576

Vzorec: C₈H₆Br₂O₃
MW: 309,94 g/mol
Bod tání: 213…217 °C
Storage Temperature: Ambient
MDL Number: MFCD00016978
CAS číslo: 2973-75-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR