4-Bromobenzaldehyde ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
A14237.22
A14237.14
A14237.36
A14237.22EA
5500
CZK
A14237.22
A14237.14
A14237.36
4-Bromobenzaldehyde ≥98%
4-Bromobenzaldehyde
Purity: 98+%
CAS No.: 1122-91-4
Molecular Formula: C7H5BrO
Formula Weight: 185.03
Flash Point: 108°(226°F)
Odour: Characteristic
Boiling Point: 66-68°/2mm
Beilstein Reference: 507100
MDL No.: MFCD00003377
Vzorec:
C₇H₅BrO MW: 185,02 g/mol Bod varu: 66…68 °C (2 mmHg) Bod tání: 55…58 °C Hustota: 1,849 g/cm³ (20 °C) Bod vzplanutí: 108 °C (226 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003377 CAS číslo: 1122-91-4 EINECS: 214-365-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...