2-Bromobenzaldehyde ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
o-Bromobenzaldehyde
A15065.14
A15065.22
A15065.36
A15065.14EA
2270
CZK
A15065.14
A15065.22
A15065.36
2-Bromobenzaldehyde ≥98%
2-Bromobenzaldehyde
Purity: 98%
CAS No.: 6630-33-7
Molecular Formula: C7H5BrO
Formula Weight: 185.03
Flash Point: 95°(203°F)
Refractive Index: 1.5960
Odour: Characteristic
Melting Point: 18-21°
Boiling Point: 228-230°
Beilstein Reference: 636132
MDL No.: MFCD00003300
Vzorec:
C₇H₅BrO MW: 185,02 g/mol Bod varu: 228…230 °C Bod tání: 18…21 °C Hustota: 1,597 Bod vzplanutí: 95 °C (203 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003300 CAS číslo: 6630-33-7 EINECS: 229-622-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...