Tisk…

9-Methylanthracene ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A15699.03 A15699.06 A15699.14
A15699.03EA 996 CZK
A15699.03 A15699.06 A15699.14
9-Methylanthracene ≥99%
9-Methylanthracene

Purity: 99% CAS No.: 779-02-2 Molecular Formula: C15H12 Formula Weight: 192.26 Melting Point: 77-79° Boiling Point: 196-197°/16mm UN No.: 3077 MDL No.: MFCD00001261

Vzorec: C₁₅H₁₂
Bod varu: 196…197 °C (16 mmHg)
Bod tání: 77…79 °C
Hustota: 1,066
Storage Temperature: Ambient
MDL Number: MFCD00001261
CAS číslo: 779-02-2
EINECS: 212-299-7
UN: 3077
ADR: 9,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR