9-(Chloromethyl)anthracene ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A13579.03
A13579.06
A13579.14
A13579.03EA
1230
CZK
A13579.03
A13579.06
A13579.14
9-(Chloromethyl)anthracene ≥98%
9-(Chloromethyl)anthracene
Purity: 98+%
CAS No.: 24463-19-2
Molecular Formula: C15H11Cl
Formula Weight: 226.71
Melting Point: 138-142°
Beilstein Reference: 1873394
UN No.: 3261
MDL No.: MFCD00001263
Vzorec:
C₁₅H₁₁Cl Bod varu: 371 °C (1013 hPa) Bod tání: 138…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001263 CAS číslo: 24463-19-2 EINECS: 246-274-7 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...