3,3-Dimethyl-1-butyne ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
3,3-Dimethylbutyne
A14013.06
A14013.14
A14013.06EA
2780
CZK
A14013.06
A14013.14
3,3-Dimethyl-1-butyne ≥98%
3,3-Dimethyl-1-butyne
Purity: 98%
CAS No.: 917-92-0
Molecular Formula: C6H10
Formula Weight: 82.15
Storage Temperature: 2°C to 8°C
Flash Point:
Vzorec:
C₆H₁₀ MW: 82,15 g/mol Bod varu: 37…38 °C Bod tání: –78 °C Hustota: 0,671 Bod vzplanutí: <–34 °C (–29 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008852 CAS číslo: 917-92-0 EINECS: 213-035-3 UN: 3295 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...