2-Hexyne ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
B22405.09
B22405.18
B22405.09EA
1790
CZK
B22405.09
B22405.18
2-Hexyne ≥98%
2-Hexyne
Purity: 98+%
CAS No.: 764-35-2
Molecular Formula: CH3(CH2)2C≡CCH3
Formula Weight: 82.15
Flash Point: -10°(14°F)
Refractive Index: 1.4140
Odour: Characteristic
Melting Point: -90°
Boiling Point: 84-86°
Beilstein Reference: 1731542
UN No.: 3295
MDL No.: MFCD00009470
Vzorec:
C₆H₁₀ MW: 82,15 g/mol Bod varu: 84…86 °C Bod tání: –90 °C Hustota: 0,736 Bod vzplanutí: –10 °C (14 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009470 CAS číslo: 764-35-2 EINECS: 212-117-6 UN: 3295 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...