Tisk…

2-Butyne ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

A13823.06 A13823.14 A13823.22
A13823.06EA 2320 CZK
A13823.06 A13823.14 A13823.22
2-Butyne ≥98%
2-Butyne

Purity: 98% CAS No.: 503-17-3 Molecular Formula: CH3C≡CCH3 Formula Weight: 54.09 Storage Temperature: 2°C to 8°C Flash Point: -49°(-56°F) Refractive Index: 1.3920 Odour: Recognizable Melting Point: -32° Boiling Point: 24-27° Beilstein Reference: 1361340 UN No.: 3295 MDL No.: MFCD00009275

Vzorec: C₄H₆
MW: 54,09 g/mol
Bod varu: 24…27 °C
Bod tání: –32 °C
Hustota: 0,690
Bod vzplanutí: –49 °C (–56 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00009275
CAS číslo: 503-17-3
EINECS: 207-962-2
UN: 3295
ADR: 3,I

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR