Tisk…

1-Octyne ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A12440.09 A12440.14 A12440.22
A12440.09EA 1040 CZK
A12440.09 A12440.14 A12440.22
1-Octyne ≥98%
1-Octyne

Purity: 98% CAS No.: 629-05-0 Molecular Formula: C8H14 Formula Weight: 110.20 Flash Point: 17°(62°F) Physical Form: Liquid Density: 0.746 Refractive Index: 1.4160 Appearance: Colourless Melting Point: -60° Boiling Point: 123-126° Beilstein Reference: 1734494 UN No.: 3295 MDL No.: MFCD00009546

Vzorec: C₈H₁₄
Bod varu: 123…126 °C
Bod tání: –60 °C
Hustota: 0,746
Bod vzplanutí: 17 °C (62 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009546
CAS číslo: 629-05-0
EINECS: 211-069-3
UN: 3295
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR