4-Methyl-1-pentene ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
33029.09
33029.18
33029.30
33029.09EA
1440
CZK
33029.09
33029.18
33029.30
4-Methyl-1-pentene ≥98%
4-Methyl-1-pentene
Purity: 98+%
CAS No.: 691-37-2
Molecular Formula: C6H12
Formula Weight: 84.16
Flash Point: -32°(-25°F)
Physical Form: Liquid
Density: 0.664
Refractive Index: 1.3820
Appearance: Colourless
Odour: Petrol-like
Melting Point: -153°
Boiling Point: 53-54°
Beilstein Reference: 1731096
UN No.: 3295
MDL No.: MFCD00008949
Vzorec:
C₆H₁₂ MW: 84,16 g/mol Bod varu: 53…54 °C Bod tání: –153 °C Hustota: 0,664 Bod vzplanutí: –32 °C (–25 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008949 CAS číslo: 691-37-2 EINECS: 211-720-1 UN: 3295 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...