2,4,4-Trimethyl-2-pentene ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
B20773.14
B20773.22
B20773.14EA
5220
CZK
B20773.14
B20773.22
2,4,4-Trimethyl-2-pentene ≥97%
2,4,4-Trimethyl-2-pentene
Purity: 97%
CAS No.: 107-40-4
Molecular Formula: C8H16
Formula Weight: 112.22
Flash Point: -1°(29°F)
Refractive Index: 1.4160
Melting Point: -106°
Boiling Point: 103-105°
UN No.: 3295
MDL No.: MFCD00008902
Vzorec:
C₈H₁₆ Bod varu: 103…105 °C Bod tání: –106 °C Hustota: 0,720 Bod vzplanutí: –1 °C (29 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008902 CAS číslo: 107-40-4 EINECS: 203-488-5 UN: 3295 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...