2,4,4-Trimethyl-1-pentene ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
B20187.14
B20187.22
B20187.14EA
1790
CZK
B20187.14
B20187.22
2,4,4-Trimethyl-1-pentene ≥99%
2,4,4-Trimethyl-1-pentene
Purity: 99%
CAS No.: 107-39-1
Molecular Formula: C8H16
Formula Weight: 112.22
Flash Point: -6°(21°F)
Refractive Index: 1.4085
Odour: Petrol-like
Melting Point: -94°
Boiling Point: 101-102°
Beilstein Reference: 1098309
UN No.: 2050
MDL No.: MFCD00008855
Vzorec:
C₈H₁₆ MW: 112,22 g/mol Bod varu: 101…102 °C Bod tání: –94 °C Hustota: 0,715 Bod vzplanutí: –6 °C (21 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008855 CAS číslo: 107-39-1 EINECS: 203-486-4 UN: 2050 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...