Tisk…

3-Chloro-2-methylpropene ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

B22674.0B B22674.18 B22674.30 AB134061.0250
B22674.0BEA 8030 CZK
B22674.0B B22674.18 B22674.30 ABCRAB134061.0250
3-Chloro-2-methylpropene ≥98%
3-Chloro-2-methylpropene

Purity: 98% CAS No.: 563-47-3 Molecular Formula: C4H7Cl Formula Weight: 90.55 Flash Point: -12°(9°F) Refractive Index: 1.4275 Melting Point: -80° Boiling Point: 71-72° Merck Reference: 14,2148 UN No.: 2554 MDL No.: MFCD00000953

Vzorec: C₄H₇Cl
Bod varu: 71…72 °C
Bod tání: –80 °C
Hustota: 0,917
Bod vzplanutí: –12 °C (9 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000953
CAS číslo: 563-47-3
EINECS: 209-251-2
UN: 2554
ADR: 3,II
Merck Index: 14,2148

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR