Tisk…

trans-3-Hexene ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: (E)-3-Hexene, 3-Hexene

L10311.06 L10311.03
L10311.06EA 4050 CZK
L10311.06 L10311.03
trans-3-Hexene ≥98%
trans-3-Hexene

Purity: 98% CAS No.: 13269-52-8 Molecular Formula: C6H12 Formula Weight: 84.16 Flash Point: -25°(-13°F) Refractive Index: 1.3940 Odour: Mild Melting Point: -113° Boiling Point: 66-67° Beilstein Reference: 1718859 UN No.: 3295 MDL No.: MFCD00009386

Vzorec: C₆H₁₂
Bod varu: 66…67 °C
Bod tání: –113 °C
Hustota: 0,677
Bod vzplanutí: –25 °C (–13 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009386
CAS číslo: 13269-52-8
EINECS: 236-261-4
UN: 3295
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR