trans-2-Hexene ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
(E)-2-Hexene
B22289.09
B22289.88
B22289.09EA
4530
CZK
B22289.09
B22289.88
trans-2-Hexene ≥99%
trans-2-Hexene
Purity: 99%
CAS No.: 4050-45-7
Molecular Formula: C6H12
Formula Weight: 84.16
Flash Point: -20°(-4°F)
Refractive Index: 1.3940
Odour: Mild
Melting Point: -99°
Boiling Point: 68-69°
Beilstein Reference: 1719020
UN No.: 3295
MDL No.: MFCD00009473
Vzorec:
C₆H₁₂ Bod varu: 68…69 °C Bod tání: –99 °C Hustota: 0,674 Bod vzplanutí: –20 °C (–4 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009473 CAS číslo: 4050-45-7 EINECS: 223-752-3 UN: 3295 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...