1-Octadecene, Sigma-Aldrich®
Dodavatel: Merck
74740-100ML
74740-500ML
SIAL74740-100MLEA
4030
CZK
SIAL74740-100ML
SIAL74740-500ML
1-Octadecene, Sigma-Aldrich®
1-Octadecene
Vzorec:
C₁₈H₃₆ MW: 252.48 Bod varu: 315 Bod tání: 17.5 Hustota: 0.789 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009003 CAS číslo: 112-88-9 EINECS: 204-012-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...