Tisk…

1-Octadecene tech. 90%, Technical Grade

Dodavatel: Thermo Fisher Scientific

L11004.AK L11004.AU
L11004.AKEA 900 CZK
L11004.AK L11004.AU
1-Octadecene tech. 90%, Technical Grade
1-Octadecene

Purity: 90% CAS No.: 112-88-9 Molecular Formula: CH3(CH2)15CH=CH2 Formula Weight: 252.48 Flash Point: 154°(309°F) Refractive Index: 1.4440 Melting Point: 15-17° Boiling Point: 178-179°/15mm Beilstein Reference: 1761067 MDL No.: MFCD00009003

Vzorec: C₁₈H₃₆
Bod varu: 178…179 °C (15 mmHg)
Bod tání: 15…17 °C
Hustota: 0,789
Bod vzplanutí: 154 °C (309 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009003
CAS číslo: 112-88-9
EINECS: 204-012-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR