Tisk…

1-Decene ≥94%

Dodavatel: Thermo Fisher Scientific

L06800.AE L06800.AU
L06800.AEEA 851 CZK
L06800.AE L06800.AU
1-Decene ≥94%
1-Decene

Purity: 94% CAS No.: 872-05-9 Molecular Formula: C10H20 Formula Weight: 140.27 Flash Point: 49°(120°F) Refractive Index: 1.4210 Melting Point: -66° Boiling Point: 170-171° Beilstein Reference: 1737236 UN No.: 3295 MDL No.: MFCD00009577

Vzorec: C₁₀H₂₀
Bod varu: 181 °C (1013 hPa)
Bod tání: –66 °C
Hustota: 0,7408 g/cm³ (20 °C)
Bod vzplanutí: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00009577
CAS číslo: 872-05-9
EINECS: 212-819-2
UN: 3295
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR