1-Decene ≥96% remainder isomers
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
31213.22
31213.36
31213.A3
31213.22EA
1120
CZK
31213.22
31213.36
31213.A3
1-Decene ≥96% remainder isomers
1-Decene
Purity: 96%
CAS No.: 872-05-9
Molecular Formula: C10H20
Formula Weight: 140.27
Flash Point: 49°(120°F)
Physical Form: Liquid
Density: 0.741
Refractive Index: 1.4210
Appearance: Colourless
Melting Point: -66°
Boiling Point: 170-171°
Beilstein Reference: 1737236
UN No.: 3295
MDL No.: MFCD00009577
Vzorec:
C₁₀H₂₀ Bod varu: 170…171 °C Bod tání: –66 °C Hustota: 0,741 Bod vzplanutí: 49 °C (120 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009577 CAS číslo: 872-05-9 EINECS: 212-819-2 UN: 3295 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...