Tisk…

1H,1H,2H-Perfluoro-1-octene ≥99%

Dodavatel: Thermo Scientific

B20920.06 B20920.14
B20920.06EA 1390 CZK
B20920.06 B20920.14
1H,1H,2H-Perfluoro-1-octene ≥99%
1H,1H,2H-Perfluoro-1-octene

Purity: 99% CAS No.: 25291-17-2 Molecular Formula: CF3(CF2)5CH=CH2 Formula Weight: 346.09 Flash Point: 20°(202°F) Refractive Index: 1.2960 Boiling Point: 106-107° Beilstein Reference: 2060994 MDL No.: MFCD00039249

Vzorec: C₈H₃F₁₃
MW: 346,09 g/mol
Bod varu: 106…107 °C
Hustota: 1,580
Bod vzplanutí: 20 °C (202 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039249
CAS číslo: 25291-17-2
EINECS: 246-791-8
UN: 1993
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR