cis-1,4-Dichloro-2-butene, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
195707-25G
195707-100G
195707-5G
SIAL195707-25GEA
2950
CZK
SIAL195707-25G
SIAL195707-100G
SIAL195707-5G
cis-1,4-Dichloro-2-butene, Sigma-Aldrich®
cis-1,4-Dichloro-2-butene
Vzorec:
C₄H₆Cl₂ MW: 125 g/mol Bod varu: 152 °C (1013 hPa) Bod tání: –48 °C Hustota: 1,1363 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00062950 CAS číslo: 1476-11-5 EINECS: 216-021-5 UN: 3390 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...