2-Chloropropene ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
α-Methylvinyl chloride
L12374.06
L12374.14
L12374.06EA
2450
CZK
L12374.06
L12374.14
2-Chloropropene ≥99%
2-Chloropropene
Purity: 99%
CAS No.: 557-98-2
Molecular Formula: CH3CCl=CH2
Formula Weight: 76.53
Storage Temperature: 2°C to 8°C
Flash Point: -34°(-29°F)
Refractive Index: 1.3939
Odour: Recognizable
Melting Point: -138°
Boiling Point: 22-23°
Beilstein Reference: 1361376
UN No.: 2456
MDL No.: MFCD00000859
Vzorec:
C₃H₅Cl MW: 76.53 Bod varu: 22…23 °C Bod tání: –138 °C Hustota: 0.899 Bod vzplanutí: –34 °C (–29 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000859 CAS číslo: 557-98-2 EINECS: 209-187-5 UN: 2456 ADR: 3 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...