Allyl acetate ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
41353.AD
41353.AK
41353.ADEA
1330
CZK
41353.AD
41353.AK
Allyl acetate ≥99%
Allyl acetate
Purity: 99%
CAS No.: 591-87-7
Molecular Formula: CH3CO2CH2CH=CH2
Formula Weight: 100.12
Physical Form: Liquid
Density: 0.928
Refractive Index: 1.4042
Melting Point: 6°
Boiling Point: 103-104°
Merck Reference: 14,285
UN No.: 2333
MDL No.: MFCD00008721
Vzorec:
C₅H₈O₂ MW: 100,12 g/mol Bod varu: 103…104 °C Bod tání: 6 °C Hustota: 0,928 Bod vzplanutí: 6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008721 CAS číslo: 591-87-7 EINECS: 209-734-8 UN: 2333 ADR: 3,II Merck Index: 14,285 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...