Tisk…

2-Bromo-1-butene ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

B20067.06 B20067.14 B20067.03
B20067.06EA 9650 CZK
B20067.06 B20067.14 B20067.03
2-Bromo-1-butene ≥97%
2-Bromo-1-butene

Purity: 97% CAS No.: 23074-36-4 Molecular Formula: CH3CH2CBr=CH2 Formula Weight: 135.00 Storage Temperature: -30°C to -10°C

Vzorec: C₄H₇Br
MW: 135 g/mol
Bod varu: 88 °C (1013 hPa)
Bod tání: –133,4 °C
Hustota: 1,315 g/cm³ (20 °C)
MDL Number: MFCD00039178
CAS číslo: 23074-36-4
EINECS: 245-412-3
UN: 1993
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR