Tisk…

Methyl phenylacetate 99+%

Dodavatel: Thermo Fisher Scientific

Synonyms: Phenylacetic acid methyll ester

148440050 148442500 148440010
ACRO148440050EA 1030 CZK
ACRO148440050 ACRO148442500 ACRO148440010
Methyl phenylacetate 99+%
Methyl phenylacetate
Vzorec: C₉H₁₀O₂
MW: 150.18
Bod varu: 218
Bod tání: 49…50 °C
Hustota: 1.067 g/cm³
Bod vzplanutí: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00008453
CAS číslo: 101-41-7
EINECS: 202-940-9
Merck Index: 12,07422

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5060 to 1.5080 (20°C, 589 nm)
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR