2-methylbutan ≥99.0%, AnalaR NORMAPUR® analytical reagent
Dodavatel: VWR Chemicals
|
Nebezpečí
|
103614TEA
2160
CZK
103614T
103616V
2-methylbutan ≥99.0%, AnalaR NORMAPUR® analytical reagent
2-methylbutan
Vzorec:
CH₃CH₂CH(CH₃)₂ Bod varu: 28 °C (1013 hPa) Bod tání: –160 °C Hustota: 0,62 g/cm³ (20 °C) Bod vzplanutí: –51 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009338 CAS číslo: 78-78-4 EINECS: 201-142-8 UN: 1265 ADR: 3,I |
Výsledky specifikačního testu
Assay | Min. 99.0 % |
IR Spectrum | Passes test |
Readily carbonisable substances | Passes test |
Acidity | Max. 0.0002 meq/g |
Alkalinity | Max. 0.0002 meq/g |
Bromine value | Max. 0.5 |
Colouration | Max. 10 APHA |
Evaporation residue | Max. 50 ppm |
n-Pentane | Max. 1.0 % |
Sulphur compounds (as S) | Max. 50 ppm |
Water | Max. 0.03 % |
Al (Aluminium) | Max. 0.1 ppm |
Ba (Barium) | Max. 0.02 ppm |
Ca (Calcium) | Max. 0.1 ppm |
Cd (Cadmium) | Max. 0.05 ppm |
Co (Cobalt) | Max. 0.02 ppm |
Cr (Chromium) | Max. 0.02 ppm |
Cu (Copper) | Max. 0.02 ppm |
Fe (Iron) | Max. 0.2 ppm |
K (Potassium) | Max. 0.05 ppm |
Mg (Magnesium) | Max. 0.05 ppm |
Mn (Manganese) | Max. 0.02 ppm |
Na (Sodium) | Max. 0.5 ppm |
Ni (Nickel) | Max. 0.02 ppm |
Pb (Lead) | Max. 0.05 ppm |
Sr (Strontium) | Max. 0.02 ppm |
Zn (Zinc) | Max. 0.1 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...