2,2,4,4,6,8,8-Heptamethylnonane, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ100
128511-100G
128511-25G
SIAL128511-100GEA
2690
CZK
SIAL128511-100G
SIAL128511-25G
2,2,4,4,6,8,8-Heptamethylnonane, Sigma-Aldrich®
2,2,4,4,6,8,8-Heptamethylnonane
Vzorec:
C₁₆H₃₄ MW: 226.45 Storage Temperature: Ambient |
MDL Number:
MFCD00008856 CAS číslo: 4390-04-9 EINECS: 224-506-8 UN: 3295 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...