tert-Pentylamine 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
tert-Amylamine
B24639.14
B24639.22
B24639.14EA
1850
CZK
B24639.14
B24639.22
tert-Pentylamine 98%
tert-Pentylamine
Purity: 98%
CAS No.: 594-39-8
Molecular Formula: CH3CH2C(CH3)2NH2
Formula Weight: 87.17
Flash Point: -1°(30°F)
Refractive Index: 1.3995
Odour: Amine-like
Boiling Point: 76-78°
UN No.: 1106
MDL No.: MFCD00008056
Vzorec:
C₅H₁₃N MW: 87,16 g/mol Bod varu: 76…78 °C Bod tání: –105 °C Hustota: 0,746 Bod vzplanutí: –1 °C (30 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008056 CAS číslo: 594-39-8 EINECS: 209-839-9 UN: 1106 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...