Tisk…

tert-Butyl isocyanate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

144452-25G 144452-100G 144452-5G
SIAL144452-25GEA 2710 CZK
SIAL144452-25G SIAL144452-100G SIAL144452-5G
tert-Butyl isocyanate, Sigma-Aldrich®
tert-Butyl isocyanate
Vzorec: C₅H₉NO
MW: 99,13 g/mol
Bod varu: 85 °C (1013 hPa)
Bod tání: < –25 °C
Hustota: 0,8359 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002035
CAS číslo: 1609-86-5
EINECS: 216-544-9
UN: 2484
ADR: 6.1,I

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR