Tisk…

1-Iodo-3-methylbutane ≥97% stabilizovaný

Dodavatel: Thermo Fisher Scientific
Varování


Stabilization: Stabilised with copper

L05194.09 L05194.18
L05194.09EA 1390 CZK
L05194.09 L05194.18
1-Iodo-3-methylbutane ≥97% stabilizovaný
1-Iodo-3-methylbutane

Stabilized with copper


Purity: 97% CAS No.: 541-28-6 Molecular Formula: C5H11I Formula Weight: 198.05 Flash Point: 40°(104°F) Refractive Index: 1.4920 Boiling Point: 146-148° Beilstein Reference: 1730990 Merck Reference: 14,5120 UN No.: 1993 MDL No.: MFCD00039414

Vzorec: C₅H₁₁I
Bod varu: 146…148 °C
Hustota: 1,515
Bod vzplanutí: 40 °C (104 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039414
CAS číslo: 541-28-6
EINECS: 208-773-8
UN: 1993
ADR: 3,III
Merck Index: 14,5120

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR