Tisk…

3-Butylacetylacetone ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3-n-Butyl-2,4-pentanedione

A11236.09 A11236.18
A11236.09EA 5700 CZK
A11236.09 A11236.18
3-Butylacetylacetone ≥98%
3-Butylacetylacetone

Purity: 98% CAS No.: 1540-36-9 Molecular Formula: C9H16O2 Formula Weight: 156.23 Refractive Index: 1.4470 Boiling Point: 101-104°/10mm MDL No.: MFCD00026235

Vzorec: C₉H₁₆O₂
MW: 156,22 g/mol
Bod varu: 101…104 °C (10 mmHg)
Hustota: 0,927
Storage Temperature: Ambient
MDL Number: MFCD00026235
CAS číslo: 1540-36-9
EINECS: 216-274-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR