Tisk…

n-Tetradekan ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Tetradecane

30809.14 30809.22 30809.36
30809.14EA 1190 CZK
30809.14 30809.22 30809.36
n-Tetradekan ≥99%
n-Tetradekan

Purity: 99+% CAS No.: 629-59-4 Molecular Formula: C14H30 Formula Weight: 198.39 Flash Point: 99°(211°F) Physical Form: Liquid Density: 0.763 Refractive Index: 1.4290 Appearance: Colourless Odour: Mild Melting Point: 6° Boiling Point: 252-254° MDL No.: MFCD00008986

Vzorec: H₃C(CH₂)₁₂CH₃
MW: 198,39 g/mol
Bod varu: 252…254 °C
Bod tání: 6 °C
Hustota: 0,763
Bod vzplanutí: 99 °C (211 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008986
CAS číslo: 629-59-4
EINECS: 211-096-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03064a29015d1e810f1e4350a7a6cd2dfe4713fd&vid=f4838a5686fe033622495a3bf280963555654d9a&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR