Tisk…

1-Ethoxy-2,2,2-trifluoroethanol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Trifluoroacetaldehyde ethyl hemiacetal, Trifluoroacetaldehydeethylhemiacetal

M-Clarity™ quality level = MQ100

T62006-25G T62006-5G
SIALT62006-25GEA 8840 CZK
SIALT62006-25G SIALT62006-5G
1-Ethoxy-2,2,2-trifluoroethanol, Sigma-Aldrich®
1-Ethoxy-2,2,2-trifluoroethanol
Vzorec: C₄H₇F₃O₂
MW: 144,09 g/mol
MDL Number: MFCD00000441
CAS číslo: 433-27-2
EINECS: 207-087-6
UN: 1989
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR