Tisk…
Potassium cyclopentyltrifluoroborate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Cyclopentyltrifluoroboric acid potassium salt
M-Clarity™ quality level = MQ100
666017-1G 666017-5G
SIAL666017-1GEA 0 CZK
SIAL666017-1G SIAL666017-5G
Potassium cyclopentyltrifluoroborate, Sigma-Aldrich®
Potassium cyclopentyltrifluoroborate
Vzorec: C₅H₉BF₃K
MDL Number: MFCD04112712
CAS číslo: 1040745-70-7